Learn More
Saint-Gobain Silicone Versilic™ Cap
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Saint Gobain Performance Plastics™ Silicone Versilic™ Caps are particularly suited for all types of tissue culture work.
162.00 SEK - 260.00 SEK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11704578
|
Saint-Gobain
26302910 |
Diameter Bottom: 23mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11724578
|
Saint-Gobain
26303405 |
Top Diameter: 35mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11794568
|
Saint-Gobain
26302710 |
Diameter Bottom: 21mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11714578
|
Saint-Gobain
26303110 |
Diameter Bottom: 26mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
Silicone |