Läs mer
Silodosin N-Oxide, TRC
Produktkod. 30421900
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421900 25 mg
30420031 50 mg
30414757 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS46504525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2361272-05-9
Molekylformel C25H32F3N3O5
Molekylvikt (g/mol) 511.53
IUPAC-namn 1-(3-hydroxypropyl)-1-oxido-5-[(2R)-2-[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethylamino]propyl]-2,3-dihydroindol-1-ium-7-carboxamide
LEDER C[C@H](Cc1cc2CC[N+]([O-])(CCCO)c2c(c1)C(=O)N)NCCOc3ccccc3OCC(F)(F)F
Specifikationer
Kemiskt namn eller material Silodosin N-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C25H32F3N3O5/c1-17(30-8-12-35-21-5-2-3-6-22(21)36-16-25(26,27)28)13-18-14-19-7-10-31(34,9-4-11-32)23(19)20(15-18)24(29)33/h2-3,5-6,14-15,17,30,32H,4,7-13,16H2,1H3,(H2,29,33)/t17-,31?/m1/s1
Formel vikt 511.2294