Learn More
Trajan Scientific and Medical™ SilTite™ Column Connector Kit
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073205
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15268913
  • 1570.00 SEK / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
0.32 mm I.D. column
SilTite Kit 0.32 mm ID Column for Shimadzu
10 ferrules, 2 nuts