Learn More
Trajan Scientific and Medical™ SilTite™ Column Connector Kit
Ensures easy, leak-free installation for capillary columns without the use of any tools.
1570.00 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15248913
|
Trajan Scientific and Medical™
073203 |
1/16 in. column |
1570.00 SEK
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
15268913
|
Trajan Scientific and Medical™
073205 |
0.32 mm I.D. column |
1570.00 SEK
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
10 ferrules, 2 nuts |