Learn More
Trajan Scientific and Medical™ SilTite™ Column Connector Kit
Ensures easy, leak-free installation for capillary columns without the use of any tools.
1570.00 SEK
Specifications
Includes 10 ferrules, 2 nuts
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15248913
View Documents Promotion Details
Trajan Scientific and Medical™
073203
1/16 in. column
1570.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15268913
View Documents Promotion Details
Trajan Scientific and Medical™
073205
0.32 mm I.D. column
1570.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
10 ferrules, 2 nuts