Learn More
Trajan Scientific and Medical™ SilTite™ Mini Union
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand: Trajan Scientific and Medical™ 073554
Additional Details : Weight : 0.01000kg
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
Mini Union | |
Inert and Precise Glass Lined Bore | |
0.8 mm | |
Connecting Capillary Columns |