missing translation for 'onlineSavingsMsg'
Learn More

Silver on alumina, 3%-4% Ag, Thermo Scientific™

Catalog Number 15462107
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
MFCD00003397
This item is not returnable. View return policy
MFCD00003397

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Concentration or Composition (by Analyte or Components) 3% to 4% Ag
Physical Form Beads
Chemical Name or Material Silver on alumina
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'