missing translation for 'onlineSavingsMsg'
Learn More

Silver bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Catalog Number 15432767
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
189114-61-2
C2AgF6NO4S2
388.004
MFCD08457644
HSYLTRBDKXZSGS-UHFFFAOYSA-N
sliverbis trifluoromethanesulfonimide
131875416
silver;bis(trifluoromethylsulfonyl)azanide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]
This item is not returnable. View return policy
189114-61-2
C2AgF6NO4S2
388.004
MFCD08457644
HSYLTRBDKXZSGS-UHFFFAOYSA-N
sliverbis trifluoromethanesulfonimide
131875416
silver;bis(trifluoromethylsulfonyl)azanide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]

Silver bis(trifluoromethylsulfonyl)imide is used in the extraction of C5 - C8 linear alfa olefins using its silver salt to form room temperature ionic liquids. Further, it is utilized for the separation of olefins from paraffins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 249°C
Color Orange to White
Physical Form Solid
Quantity 250 mg
Linear Formula (CF3SO2)2NAg
UN Number UN1759
Solubility Information Soluble in water.
Formula Weight 388
Sensitivity Light and moisture sensitive
Chemical Name or Material Silver bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'