Learn More
Silver cyanide, 99%, for analysis, Thermo Scientific Chemicals
Catalog Number p-221900
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
506-64-9
CAgN
133.89
MFCD00003409
LFAGQMCIGQNPJG-UHFFFAOYSA-N
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent
10475
silver;cyanide
[C-]#N.[Ag+]
506-64-9
CAgN
133.89
MFCD00003409
LFAGQMCIGQNPJG-UHFFFAOYSA-N
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent
10475
silver;cyanide
[C-]#N.[Ag+]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 98.5% min
Linear Formula AgCN
Fieser 01,1006; 05,581; 07,320
Merck Index 15, 8651
Solubility Information Solubility in water: insoluble. Other solubilities: sol. in nh4oh,alkali cyanides,boiling conc. hno3,insoluble in alcohol and diluted acids
Formula Weight 133.89
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Silver cyanide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P361-Take off immediately all contaminated clothing.