Learn More
MP Biomedicals™ Silver Enhancing Kit, 2x250mL
Used for amplifying the signal from gold particles attached to proteins or antigens immobilized on membranes
Brand: MP Biomedicals™ 08678211
Additional Details : Weight : 0.01000kg
Specifications
Silver Enhancing Kit | |
Immunoblotting, Immunoassays | |
2 x 250 mL | |
2° to 8°C |