missing translation for 'onlineSavingsMsg'
Learn More
Silver Nitrate Solution 0.05M (0.05N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10746101

Silver Nitrate Solution 0.05M (0.05N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. p-7073177 Shop All Fisher Chemical Products
1 L, 1 L
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
This item is not returnable. View return policy

Product Code. 10746101

Brand: Fisher Chemical J/7340/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
IUPAC Name silver;nitrate
Packaging HDPE plastic bottle
Quantity 1 L
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number2540284 Certificate TypeCertificate of Analysis Date09/06/2025 Product CodeJ/7340/15,J/7340/17
Lot Number2530417 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/7340/15,J/7340/17
compliance-icons
Product Identifier
  • Silver nitrate 0.05M
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver Nitrate Solution 0.05M (0.05N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.