missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver p-toluenesulfonate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.
Solubility
Soluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.
Solubility
Soluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Quantity | 5 g |
UN Number | UN3077 |
Solubility Information | Soluble in water. |
Formula Weight | 279.07 |
Percent Purity | ≥98% |
Odor | Odorless |
Sensitivity | Light sensitive |
Chemical Name or Material | Silver p-toluenesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'