Learn More
Silver Trifluoromethanesulfonate, Sigma-Aldrich™
Product Code. 15899880
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
15899880 5 g 5g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15899880 Supplier Sigma Aldrich Supplier No. 8143250005
 Restricted Item
Beräknad leverans: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This catalyst efficiently promotes decarboxylative functionalization of alkynoic acids. Its ability to generate cationic gold species makes it invaluable for tandem catalysis in complex molecule synthesis.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2923-28-6
Melting Point 356°C
MDL Number MFCD00013226
Quantity 5 g
Synonym Silver trifluoromethanesulfonate, Trifluoromethanesulfonic acid silver salt, Silver trifluoromethanesulfonate
Molecular Weight (g/mol) 256.94
Grade Research