Läs mer
Silybin A, TRC
Produktkod. 30412053
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30412053 1 mg
1 options
Produktkod. 30412053 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS4658551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Silybin A
CAS 22888-70-6
Molekylformel C25 H22 O10
Kvantitet 1 mg
Synonym Silybin, (2R,3R)-2-[(2R,3R)-2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7-Trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxan-6-yl]-4-chromanone, [2R-[2α,3β,6(2R*,3R*)]]-2-[2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 1,4-Benzodioxin 4H-1-benzopyran-4-one deriv., 7C3MT, Silibinin, Silibinin A, Silliver, Silybin A, Silybin b1, Silybine, Silybum substance E6, Silymarin I, Silymarin MZ 80, Silymarine I
InChI formel InChI=1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20-,23+,24-,25-/m1/s1
LEDER COc1cc(ccc1O)[C@H]2Oc3cc(ccc3O[C@@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O
IUPAC-namn (2R,3R)-3,5,7-trihydroxy-2-[(2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-2,3-dihydrochromen-4-one
Molekylvikt (g/mol) 482.44
Formel vikt 482.1213