Läs mer
Silymarin (>80%), TRC
Produktkod. 30132933
Change view
Kvantitet:
300 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132933 25 g
30132736 300 mg
17982855 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132933 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS46589525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65666-07-1
Molekylformel C25H22O10
Molekylvikt (g/mol) 482.44
Synonym Apihepar, Darsil, Flavobion, Karsilin, Laragon, Legalon, Legalon 70, Pluropon, Silarin, Silepar, Silirex, Silistrong, Silmar, Silymarin Group, Silystrong
IUPAC-namn 3,5,7-trihydroxy-2-(3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one
LEDER COC1=C(O)C=CC(C2C(CO)OC3=C(C=C(C4C(O)C(C5=C(O)C=C(O)C=C5O4)=O)C=C3)O2)=C1
Specifikationer
Kemiskt namn eller material Silymarin
Kvantitet 25 g
InChI formel InChI=1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3
Formel vikt 482.12