Learn More
Simport™ Scientific T310 Series Cryovials
Designed for storing biological material, human or animal cells, these cryovials are compatible with most storage systems
Brand:  Simport™ Scientific T310-10A
Additional Details : Weight : 2.70000kg
Product Code. 11640801
  • 3431.00 SEK / Pack of 500
In Stock
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Cap features a long skirt for easy one-hand aseptic methods and an exclusive silicone seal fitted inside the cap
  • Super-fast thread design allows removal or sealing with a mere 1™1/4 turn
  • Inside thread design that will not contribute to possible contamination
  • Tubes are provided with a white marking area for sample identification
  • Sterilized by gamma radiation
  • Autoclavable
  • Polypropylene construction
Specifications
Specifications
Polypropylene
10 mL
External
Yes
Screw Cap
Yes
Sterile
For storing biological material, human or animal cells, these cryovials are compatible with most storage systems
Yes
Autoclavable
500/Cs.