Learn More
Simport™ Scientific T310 Series Cryovials
T310 Series Cryovials are designed for storing biological material, human or animal cells, these cryovials are compatible with most storage systems.
POA
Specifications
Material | Polypropylene |
---|---|
Sterility | Sterile |
For Use With (Application) | For storing biological material, human or animal cells, these cryovials are compatible with most storage systems |
Thread Style | External |
RNase Free | Yes |
Description
- Cap features a long skirt for easy one-hand aseptic methods and an exclusive silicone seal fitted inside the cap
- Super-fast thread design allows removal or sealing with a mere 1 1/4 turn
- Inside thread design that will not contribute to possible contamination
- Tubes are provided with a white marking area for sample identification
- Sterilized by gamma radiation
- Autoclavable
- Polypropylene construction
Specifications
Polypropylene | |
For storing biological material, human or animal cells, these cryovials are compatible with most storage systems | |
Yes | |
Autoclavable | |
Yes |
Sterile | |
External | |
Yes | |
Cryogenic Vial |