Läs mer
Simvastatin Acyl-Beta-D-glucuronide, TRC
Produktkod. 30416516
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30416516 5 mg
1 options
Produktkod. 30416516 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS4850055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Simvastatin Acyl-beta-D-glucuronide
CAS 463962-56-3
Molekylformel C31 H48 O12
Kvantitet 5 mg
Synonym Simvastatin Acyl-B-D-Glucuronide
InChI formel InChI=1S/C31H48O12/c1-6-31(4,5)30(40)41-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(32)13-19(33)14-22(34)42-29-26(37)24(35)25(36)27(43-29)28(38)39/h7-8,11,15-16,18-21,23-27,29,32-33,35-37H,6,9-10,12-14H2,1-5H3,(H,38,39)/t15-,16-,18+,19+,20-,21-,23-,24-,25-,26+,27-,29+/m0/s1
LEDER CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)[C@@H]12
IUPAC-namn (2S,3S,4S,5R,6S)-6-[(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 612.71
Formel vikt 612.3146