Läs mer
Sinapine Thiocyanate, TRC
Produktkod. 30422480
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30422480 100 mg
1 options
Produktkod. 30422480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS486810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sinapine Thiocyanate
CAS 7431-77-8
Molekylformel C16 H24 N O5 . C N S
Kvantitet 100 mg
Synonym 4-Hydroxy-3,5-dimethoxycinnamatecholine Thiocyanate, 2-[[3-(4-Hydroxy-3,5-dimethoxyphenyl)-1-oxo-2-propenyl]oxy]-N,N,N-trimethyllethanaminium Thiocyanate
InChI formel InChI=1S/C16H23NO5.CHNS/c1-17(2,3)8-9-22-15(18)7-6-12-10-13(20-4)16(19)14(11-12)21-5;2-1-3/h6-7,10-11H,8-9H2,1-5H3;3H
LEDER COc1cc(\C=C\C(=O)OCC[N+](C)(C)C)cc(OC)c1O.[S-]C#N
IUPAC-namn 2-[(E)-3-(4-hydroxy-3,5-dimethoxy-phenyl)prop-2-enoyl]oxyethyl-trimethyl-ammonium;thiocyanate
Molekylvikt (g/mol) 368.45
Formel vikt 368.1406