Learn More
R&D Systems™ Single Cell Cartridge Kit for Pala Systems (10 Cartridges)
Descrizione
- Sterile, single-use microfluidic cartridges designed for precise single-cell isolation and dispensing
- Enables deposition of one viable cell per well into 96- and 384-well plates for clonal expansion
- Supports high-efficiency monoclonal cell line development and single-cell genomics workflows
- Microfluidic design ensures gentle handling to preserve viability of sensitive and primary cells
- Individually packaged cartridges minimize contamination risk and support sterile workflows
- Optimized for samples with typical concentrations of 5,000 to20,000 cells/mL
- Processes sample volumes up to approximately 600 μL
- Provides reliable single-cell dispensing accuracy for downstream analytical confidence
- Disposable format eliminates cleaning requirements and reduces system downtime
- Compatible with R&D Systems™ Pala and Hana Single Cell Dispenser systems
- Supports applications including monoclonal cell line generation, CRISPR workflows, antibody discovery, and single-cell analysis
- Kit includes 10 cartridges for consistent, repeatable experimental workflows
Specifica
Specifica
| Product Type | Cartridge |
| For Use With (Equipment) | Pala Single Cell Sorter and Dispenser |
| Includes | 10 Sterile Single-use Microfluidic Cartridges |
| Product Line | ProteinSimple |
Designed for use with Pala systems. Cartridges are sterile and individually packaged for single-use workflows. Performance may vary depending on sample preparation and cell characteristics.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?