Learn More
Epredia™ Single Cytofunnel™ with White Filter Cards
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
Brand: Epredia™ 5991040
Additional Details : Weight : 0.54000kg
Description
- Disposable sample chambers with white filter cards and caps
- Pre-attached filter card
Specifications
White | |
50 Pk. | |
0.1 to 0.5 mL |
Single-use | |
Single Cytofunnel™, With Filter Cards and Caps |