Läs mer
Axygen™ Enkelbrunns högprofilsreagensbehållare
Handla allt Axygen Scientific Produkter
Kapacitet (metrisk):
240 mL
282 mL
290 mL
Sterilitet:
Icke-steril
Non-sterile
Sterila
Kanaler:
8
12
96
384
Förpackningsstorlek:
Paket med 25
Beskrivning
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifikationer
Specifikationer
| Certifikat och efterlevnad | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
| Färg | Clear |
| Kapacitet (metrisk) | 282 mL |
| Material | Polypropylene |
| Sterilitet | Non-sterile |
| Typ | Reagent Reservoir |
| Förpackning | 25/Cs. |
| Kanaler | 384 |
| Disponibel | Single-use |