Läs mer
Sinigrin Monohydrate, TRC
Produktkod. 30409217
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409217 5 mg
30410161 50 mg
30409440 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30409217 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS4879135MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64550-88-5
Molekylformel C10H16KNO9S2.H2O
Molekylvikt (g/mol) 397.5
Synonym β-D-Glucopyranose, 1-thio-, 1-[N-(sulfooxy)-3-butenimidate], monopotassium salt, monohydrate, (Z)- (9CI), Sinigrine potassium monohydrate, 1-Thio-β-D-glucopyranose (Z)-1-[N-(sulfooxy)-3-butenimidate] monopotassium salt monohydrate
IUPAC-namn potassium;[(E)-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanylbut-3-enylideneamino] sulfate;hydrate
LEDER O.[K+].OC[C@H]1O[C@@H](S\C(=N/OS(=O)(=O)[O-])\CC=C)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Sinigrin Monohydrate
Kvantitet 5 mg
InChI formel InChI=1S/C10H17NO9S2.K.H2O/c1-2-3-6(11-20-22(16,17)18)21-10-9(15)8(14)7(13)5(4-12)19-10;;/h2,5,7-10,12-15H,1,3-4H2,(H,16,17,18);;1H2/q;+1;/p-1/b11-6+;;/t5-,7-,8+,9-,10+;;/m1../s1
Formel vikt 415.0009