Läs mer
Sinigrine, TRC
Produktkod. 30417081
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30417081 50 mg
1 options
Produktkod. 30417081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS48735050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sinigrin Potassium salt
CAS 3952-98-5
Molekylformel C10 H16 N O9 S2 . K
Kvantitet 50 mg
Synonym β-D-Glucopyranose','1-thio-','1-[N-(sulfooxy)-3-butenimidate]','potassium salt (1:1) (ACI), Glucopyranose','1-thio-','1-(3-butenoate)','oxime','O-sulfate','potassium salt','β-D- (7CI), Glucopyranose','1-thio-','1-(3-butenohydroximate) NO-(hydrogen sulfate)','monopotassium salt','β-D- (8CI), Sinigrin (6CI), β-D-Glucopyranose','1-thio-','1-[N-(sulfooxy)-3-butenimidate]','monopotassium salt (9CI), Allyl glucosinolate potassium, NSC 407279, NSC 90774, Sinigrine
InChI formel InChI=1S/C10H17NO9S2.K/c1-2-3-6(11-20-22(16,17)18)21-10-9(15)8(14)7(13)5(4-12)19-10;/h2,5,7-10,12-15H,1,3-4H2,(H,16,17,18);/q;+1/p-1/b11-6+;/t5-,7-,8+,9-,10+;/m1./s1
LEDER [K+].OC[C@H]1O[C@@H](S\C(=N\OS(=O)(=O)[O-])\CC=C)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn potassium;[(E)-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanylbut-3-enylideneamino] sulfate
Molekylvikt (g/mol) 397.46
Formel vikt 396.9904