missing translation for 'onlineSavingsMsg'
Learn More

CX3CL1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human CX3CL1 (NP_002987.1) extracellular domain (Met 1-Arg 339) fused to a C-terminal polyhistidine tag

Brand:  Invitrogen 10636H08H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247005

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Description

Description

Chemokine (C-X3-C Motif) Ligand 1 (CX3CL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to compete with biotinylated recombinant human CX3CL1-His for binding to immobilized recombinant mouse CD4-his (Mfr. No. 50134-M08) in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6376
PAGE
5 x 5μg
C-Terminal
Other Cytokines
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
CX3CL1
Chemokines & Receptors
His Tag
Lyophilized
35kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Chemokine (C-X3-C motif) ligand 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.