Learn More
GPC5 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) fused to the polyhistidine tag at the C-terminus
Brand: Invitrogen 10079H08B25
Additional Details : Weight : 0.10000kg
Description
Glypican 5 (GPC5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC5 at 5μg/mL (100μL/well) can bind human FGF2 with a linear range of 0.156-2.5ng/mL
- Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5 x 5μg | |
<1 EU/μg | |
Glypican 5 | |
His Tag | |
Other Proteins | |
Proteoglycans |
60.5kDa | |
Store at -80 °C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
GPC5, bA93M14.1, glypican proteoglycan 5, glypican-5 | |
GPC5 | |
Insect | |
Frozen | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.