missing translation for 'onlineSavingsMsg'
Learn More

HTRA2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of human HTRA2 (O43464-1) (Ala 134-Glu 458) fused to a polyhistide tag at the C-terminus

POA

Specifications

Purity ≥85% by SDS-PAGE
Format Frozen
Gene Alias OMI, PARK13, PRSS25
View More Specs

Description

Description

HTRA Serine Peptidase 2 (HTRA2), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Protease activity demonstrated by HTRA2 cleavage of bovine beta-casein (Sigma Cat. No. C-6905); incubation of beta-casein at 0.2μg/mL with Recombinant Human HTRA2 at 0.02μg/mL (ratio of 10:1) for 60 minutes at 45°C in 50mM Tris, pH 8.0, which results in >95% cleavage of beta-casein, as revealed by SDS-PAGE
  • Formulation: 50mM Tris, 0.3M NaCl, 1mM DTT, 0.2μg/μL BSA, 20% glycerol, pH 7.8
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Frozen
OMI, PARK13, PRSS25
SDS

For Research Use Only. Not for use in diagnostic procedures.