missing translation for 'onlineSavingsMsg'
Learn More

IGFBP7 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human IGFBP7 (Q16270) (Asp 30-Leu 282) fused to a polyhistidine tag at the N-terminus

Brand:  Invitrogen 13100H07H250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246165

  • 17809.20 SEK / 250µg


Estimated Shipment date
from Supplier 06-03-2019
Add to basket

Description

Description

Insulin-like growth factor binding protein 7 (IGFBP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind biotinylated recombinant human CCL21 in a functional ELISA; 2. Measured by its ability to bind biotinylated recombinant human VEGF165 in a functional ELISA; 3. Measured by its ability to bind biotinylated recombinant human IGF2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28.6kDa
5 x 50μg
IGFs (Insulin-Like Growth Factors)|Growth Factor Receptors
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF,TAF
IGFBP7
Growth Factors & Receptors
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Insulin-like growth factor binding protein 7
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.