missing translation for 'onlineSavingsMsg'
Läs mer

IL6 Recombinant Human Protein, Invitrogen™ Sino Biological™

Produktkod. 13463009
Klicka för att se tillgängliga alternativ
Kvantitet:
5 μg
5 x 5 μg
50 μg
Förpackningsstorlek:
50µg
5µg
Paket med 5
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 13463009

Brand: Invitrogen 10395HNAE50

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This recombinant protein is expressed from a DNA sequence encoding the mature form of human IL6 (NP_000591.1) (Val 30-Met 212) retaining the initial Met at the N-terminus

Interleukin 6 (IL-6) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.&line-break;Background: Interleukin-6 (IL-6) is a multifunctional α-helical cytokine that regulates cell growth and differentiation of various tissues, and is known particularly for its role in the immune response and acute phase reactions. IL-6 is also involved in hematopoiesis, bone metabolism, and cancer progression, and has been defined an essential role in directing transition from innate to acquired immunity. IL-6 is secreted by a variety of cell types including T cells and macrophages as phosphorylated and variably glycosylated molecule with the molecular mass of 22-28kDa. Three IL-6 signaling pathways (STAT1, STAT3, and Ras-dependent MAPK cascade) have been reported. Dysregulated production of IL-6 and IL6R are implicated in the pathogenesis of several inflammatory diseases and malignancies such as multiple myeloma, rheumatoid arthritis, or osteoporosis.
  • N-terminal Sequence Analysis: Met
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 (100μL/well of a 8μg/mL solution) has been shown to bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL; 2. Measured in a cell proliferation assay using TF1 human erythroleukemic cells, the EC50 for this effect is typically 0.2-0.8ng/mL
  • Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).

TRUSTED_SUSTAINABILITY

Specifikationer

För användning med (applikation) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Molekylvikt (g/mol) 20.3kDa
Kvantitet 50 μg
Förvaringskrav Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Strukturell form Full Length/Recombinant
Gene Alias BSF2, HGF, HSF, IFNB2, IL-6
Vanligt namn Interleukin 6 (interferon, beta 2)
Gensymbol IL6
Protein Tag Untagged
Uttryckssystem E. coli
Proteinfamilj Cytokines & Receptors
Form Lyophilized
Protein subtyp Interleukins
Renhet eller kvalitetsklass ≥95% by SDS-PAGE
Visa mer Visa mindre

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.