Learn More
mMP1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the pro form of humanmMP1 enzyme (NP_002412.1) precursor (Met 1-Asn 469) fused to a C-terminal polyhistidine tag
Brand: Invitrogen 10532H08H50
Additional Details : Weight : 0.10000kg
Description
Matrix Metallopeptidase 1 (MMP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (AnaSpec Cat. No. 27076), the specific activity is >70pmole/min./μg
- Formulation: 25mM MES, 10mM CaCl2, 150mM NaCl, 0.05% BRIG 35, pH 5.5
- Reconstitution: No reconstitution needed
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50μg | |
<1 EU/μg | |
Matrix Metallopeptidase 1 | |
His Tag | |
Other Proteins | |
Proteases |
52kDa | |
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
CLG, CLGN | |
MMP1 | |
Mammalian | |
Frozen | |
≥95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.