Learn More
OLR1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human OLR1 (P78380) extracellular domain (Ser 61-Gln 273) fused to a polyhistidine tag at the N-terminus
Brand:  Invitrogen 10585H07H250
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15236965
  • 8557.80 SEK / 250µg
Estimated Shipment date
from Supplier 10-05-2019
Add to basket
Description
Description
Oxidized Low Density Lipoprotein (Lectin-like) Receptor 1 (OLR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4973
PAGE
5 x 50μg
N-Terminal
<1 EU/μg
Oxidized low density lipoprotein (lectin-like) receptor 1
Assay Standard
Mammalian
Lyophilized
26.5kDa
>85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
OLR1, CLEC8A, LOX1, LOXIN, SCARE1, SLOX1
OLR1
Other Proteins
His Tag