Learn More
RETSCH Sintered Aluminum Oxide Aeration Lids
24895.14 SEK - 37905.20 SEK
Specifications
Product Type | Aeration Lid |
---|---|
Closure Type | Aeration Lid |
Material | Sintered Aluminum Oxide |
Product Code | Brand | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Equipment) | Price | Quantity & Availability | ||||
11974372
|
RETSCH
22.107.0014 |
Sintered Aluminum Oxide | For 250 mL Comfort Grinding Jar |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11904382
|
RETSCH
22.107.0013 |
Sintered Aluminum Oxide | For 500 mL Comfort Grinding Jar |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Aeration Lid | |
Aeration Lid | |
Sintered Aluminum Oxide |