Läs mer
RETSCH Sintered Corundum Comfort Grinding Jars
Produktkod. 10398032 Handla allt Retsch Produkter
Kapacitet (metrisk):
50 mL
125 mL
250 mL
500 mL
Förpackningsstorlek:
Varje
Produktkod. 10398032
Brand: RETSCH 01.462.0226
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Use these grinding jars with Retsch™ ball mills. Retsch™ Sintered Corundum Comfort Grinding Jars feature an O-type sealing for safe operation.
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifikationer
Produkttyp Comfort Grinding Jar
Kapacitet (metrisk) 500 mL
För användning med (utrustning) For PM 100, PM 400 and S 100
Material Sintered Corundum