Läs mer
Siponimod, TRC
Produktkod. 30416140
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30416140 2.5 mg 2.5mg
30414380 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6836602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Siponimod
CAS 1230487-00-9
Molekylformel C29H35F3N2O3
Kvantitet 2.5 mg
Synonym 1-[[4-[(1E)-1-[[[4-Cyclohexyl-3-(trifluoromethyl)phenyl]methoxy]imino]ethyl]-2-ethylphenyl]methyl]-3-azetidinecarboxylic Acid, 1-(4-[1-[(E)-4-Cyclohexyl-3-trifluoromethyl-benzyloxyimino]-ethyl]-2-ethyl-benzyl)-azetidine-3-carboxylic Acid, BAF 312
InChI formel InChI=1S/C29H35F3N2O3/c1-3-21-14-23(10-11-24(21)15-34-16-25(17-34)28(35)36)19(2)33-37-18-20-9-12-26(22-7-5-4-6-8-22)27(13-20)29(30,31)32/h9-14,22,25H,3-8,15-18H2,1-2H3,(H,35,36)/b33-19+
LEDER CCc1cc(ccc1CN2CC(C2)C(=O)O)\C(=N\OCc3ccc(C4CCCCC4)c(c3)C(F)(F)F)\C
IUPAC-namn 1-[[4-[(E)-N-[[4-cyclohexyl-3-(trifluoromethyl)phenyl]methoxy]-C-methylcarbonimidoyl]-2-ethylphenyl]methyl]azetidine-3-carboxylic acid
Molekylvikt (g/mol) 516.6
Formel vikt 516.26