Läs mer
Sisapronil, TRC
Produktkod. 30136117
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136117 25 mg -
30007727 2.5 mg 2.5mg
30009393 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136117 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS12585025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856225-89-3
Molekylformel C15 H6 Cl2 F8 N4
Molekylvikt (g/mol) 465.13
Synonym 1H-Pyrazole-3-carbonitrile, 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[2,2-difluoro-1-(trifluoromethyl)cyclopropyl]-, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[2,2-difluoro-1-(trifluoromethyl)cyclopropyl]-1H-pyrazole-3-carbonitrile, PF 0241851, Sisapronil
IUPAC-namn 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[2,2-difluoro-1-(trifluoromethyl)cyclopropyl]pyrazole-3-carbonitrile
LEDER Nc1c(c(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)C3(CC3(F)F)C(F)(F)F
Specifikationer
Kemiskt namn eller material Sisapronil
Kvantitet 25 mg
InChI formel InChI=1S/C15H6Cl2F8N4/c16-6-1-5(14(20,21)22)2-7(17)10(6)29-11(27)9(8(3-26)28-29)12(15(23,24)25)4-13(12,18)19/h1-2H,4,27H2
Formel vikt 463.9842