Läs mer
Sisomicin, TRC
Produktkod. 30138542
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138542 1 g
30136749 50 mg
30008582 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138542 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS4885001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32385-11-8
Molekylformel C19 H37 N5 O7
Molekylvikt (g/mol) 447.53
Synonym D-Streptamine, O-3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-α-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-, Sisomicin (8CI), O-3-Deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-α-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-D-streptamine, Antibiotic 66-40, Antibiotic 6640, Rickamicin, Sch 13475, Siseptin
IUPAC-namn (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol
LEDER CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3OC(=CC[C@H]3N)CN)[C@@H]2O)OC[C@]1(C)O
Specifikationer
Kemiskt namn eller material Sisomicin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C19H37N5O7/c1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17/h3,9-18,24-27H,4-7,20-23H2,1-2H3/t9-,10+,11-,12+,13-,14-,15+,16-,17-,18-,19+/m1/s1
Formel vikt 447.2693
Procent renhet >95%