Läs mer
Sitafloxacin Sesquihydrate, TRC
Produktkod. 30135570
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135570 50 mg
30004719 10 mg
30009904 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS49092050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163253-35-8
Molekylformel 2 C19 H18 Cl F2 N3 O3 . 3 H2 O
Molekylvikt (g/mol) 873.67
Synonym 3-Quinolinecarboxylic acid, 7-[(7S)-7-amino-5-azaspiro[2.4]hept-5-yl]-8-chloro-6-fluoro-1-[(1R,2S)-2-fluorocyclopropyl]-1,4-dihydro-4-oxo-, hydrate (2:3), 3-Quinolinecarboxylic acid, 7-(7-amino-5-azaspiro[2.4]hept-5-yl)-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-1,4-dihydro-4-oxo-, hydrate (2:3), [1R-[1α(S*),2α]]-, Sitafloxacin sesquihydrate
IUPAC-namn 7-[(7S)-7-amino-5-azaspiro[2.4]heptan-5-yl]-8-chloro-6-fluoro-1-[(1R,2S)-2-fluorocyclopropyl]-4-oxoquinoline-3-carboxylic acid;trihydrate
LEDER O.O.O.N[C@@H]1CN(CC12CC2)c3c(F)cc4C(=O)C(=CN([C@@H]5C[C@@H]5F)c4c3Cl)C(=O)O.N[C@@H]6CN(CC67CC7)c8c(F)cc9C(=O)C(=CN([C@@H]%10C[C@@H]%10F)c9c8Cl)C(=O)O
Specifikationer
Kemiskt namn eller material Sitafloxacin Sesquihydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/2C19H18ClF2N3O3.3H2O/c2*20-14-15-8(17(26)9(18(27)28)5-25(15)12-4-10(12)21)3-11(22)16(14)24-6-13(23)19(7-24)1-2-19;;;/h2*3,5,10,12-13H,1-2,4,6-7,23H2,(H,27,28);3*1H2/t2*10-,12+,13+;;;/m00.../s1
Formel vikt 872.2326
Procent renhet >95%