Learn More
SIX1 Monoclonal Antibody (CL0185), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524588
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CFKEKSRGVL REWYAHNPYP SPREKRELAE ATGLTTTQVS NWFKNRRQRD RAAEAKEREN TENNNSSSNK QNQLSPLEGG KPLMSSSEEE FSPPQSPDQN SVLLLQGNMG HARSSNYSLP GLTASQPSHG LQTHQHQLQD S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
SIX1 Monoclonal antibody specifically detects SIX1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SIX1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15475 | |
SIX1 | |
Recombinant protein corresponding to Human SIX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
CL0185 | |
Unconjugated | |
SIX1 | |
BB138287; BOS3; DFNA23; Homeobox protein SIX1; sine oculis homeobox homolog 1; sine oculis-related homeobox 1; sine oculis-related homeobox 1 homolog; SIX homeobox 1; SIX1; TIP39 | |
Mouse | |
Protein A | |
RUO | |
6495 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only