missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIX2 Mouse anti-Canine, Human, Mouse, Porcine, Rat, Clone: 1F9G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66347-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.Specifications
SIX2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62232, Q9NPC8 | |
Six2 | |
SIX2 Fusion Protein Ag2124 | |
150 μL | |
Primary | |
Canine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1F9G11 | |
Unconjugated | |
SIX2 | |
Homeobox protein SIX2, Sine oculis homeobox homolog 2, SIX homeobox 2, SIX2 | |
Mouse | |
Protein A | |
RUO | |
100525814, 10736, 20472, 366542, 481357 | |
-20°C | |
Liquid |