missing translation for 'onlineSavingsMsg'
Learn More

SIX2 Mouse anti-Canine, Human, Mouse, Porcine, Rat, Clone: 1F9G11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66347-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888135

  • 1656.73 SEK / 20µL
Estimated Shipment: 02-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
Specifications

Specifications

SIX2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62232, Q9NPC8
Six2
SIX2 Fusion Protein Ag2124
20 μL
Primary
Canine, Mouse, Porcine, Rat, Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
1F9G11
Unconjugated
SIX2
Homeobox protein SIX2, Sine oculis homeobox homolog 2, SIX homeobox 2, SIX2
Mouse
Protein A
RUO
100525814, 10736, 20472, 366542, 481357
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers