Läs mer
SJB3-019A, MedChemExpress
Produktkod. 16487966
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487966 100 mg
16438248 10 mg
16420698 5 mg
16414137 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16487966 Leverantör Medchem Express Leverantörsnummer HY80012
MedChemExpress SJB3-019A is a potent and novel USP1 inhibitor, 5 times more potent than SJB2-043 in promoting ID1 degradation and cytoxicity in K562 cells with IC50 of 0.0781 μM.
SJB3-019A is a potent and novel USP1 inhibitor, 5 times more potent than SJB2-043 in promoting ID1 degradation and cytoxicity in K562 cells with IC50 of 0.0781 μM.
Specifikationer
Kemiskt namn eller material SJB3-019A
Fysisk form Solid
CAS 2070015-29-9
Färg Light Yellow
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H8N2O3
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 10 mg/mL (36.20 mM; Need ultrasonic)
LEDER O=C1C2=C(OC(C3=CC=CN=C3)=N2)C(C4=CC=CC=C41)=O
Molekylvikt (g/mol) 276.25
Formel vikt 276.25
Procent renhet 98.0%