missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ SJF 1528
GREENER_CHOICE

Produktkod. 18799686
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18799686 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799686 Supplier Tocris Bioscience™ Supplier No. 7262/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent EGFR PROTAC™; also degrades HER2

SJF 1528 is a potent EGFR PROTAC™ Degrader (DC50 values are 39.2 nM for wild-type EGFR in OVCAR8 cells and 736 nM for Exon20Ins mutated EGFR in HeLa cells). Also degrades HER2. SJF 1528 comprises the EGFR inhibitor lapatinib (Cat. No. 6811) joined by a linker to a von Hippel-Lindau (VHL) recruiting ligand. Inhibits proliferation of HER2-driven breast cancer cell lines (IC50 = 102 nM for SKBr3 cells). PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (2 S,4 R)-1-(-2-(2-(2-(2-(4-(4-((3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6- yl)phenoxy)ethoxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2230821-38-0
Kvantitet 5 mg
Mål Active Degraders
Molekylformel C55H57ClFN7O8S
Renhet 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.