Läs mer
Tocris Bioscience™ SJPYT 310
Beskrivning
SJPYT 310 is a potent and selective PXR antagonist (IC50 values are 4 and 9 nM for binding and cellular activity, respectively). SJPYT 310 displays selectivity for hPXR without affecting the activity of mPXR, hCAR, hFXR, hLXRα, or hVDR. SJPYT 310 shows no toxicity.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[5-(1,1-Dimethylethyl)-2-[(1S)-1-methylbutoxy]phenyl]-1-(2-methoxy-5-methylphenyl)-5-methyl-1H-1,2,3-triazole-4-carboxamide |
| CAS | 2883814-84-2 |
| Kvantitet | 10 mg |
| Mål | PXR/NR1I2 |
| Molekylformel | C27H36N4O3 |
| Renhet | 98% |