Läs mer
SKF-82958 hydrobromide, MedChemExpress
Produktkod. 16494976
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16494976 100 mg
16493968 10 mg
16440327 50 mg
16436687 5 mg
16402936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16494976 Leverantör Medchem Express Leverantörsnummer HY10435A
MedChemExpress SKF-82958 ((±)-SKF 82958) hydrobromide is a dopamine D1 receptor full agonist (K0.5=4 nM), displays selective for D1 over D2 receptors (K0.5=73 nM). SKF-82958 hydrobromide induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50=491 nM).
SKF-82958 ((±)-SKF 82958) hydrobromide is a dopamine D1 receptor full agonist (K0.5=4 nM), displays selective for D1 over D2 receptors (K0.5=73 nM). SKF-82958 hydrobromide induces dopamine D1 receptor-dependent adenylate cyclase activity in rat striatal membranes (EC50=491 nM).
Specifikationer
Kemiskt namn eller material SKF-82958 hydrobromide
CAS 74115-01-8
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C19H21BrClNO2
Kvantitet 100 mg
Synonym (±)-SKF-82958 hydrobromideChloro-APB hydrobromide
Löslighetsinformation DMSO : ≥ 100 mg/mL (243.47 mM)
LEDER OC1=C(O)C=C2C(C3=CC=CC=C3)CN(CC=C)CCC2=C1Cl.[H]Br
Molekylvikt (g/mol) 410.73
Formel vikt 410.73
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation