missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKIV2L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12719-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SKIV2L2 gene ontology annotations related to this gene include RNA catabolic process.Specifications
SKIV2L2 | |
Polyclonal | |
Unconjugated | |
SKIV2L2 | |
ATP dependent helicase SKIV2L2, Dob1, fSAP118, KIAA0052, Mtr4, SKIV2L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23517, 365668, 72198 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42285, Q9CZU3 | |
Skiv2l2 | |
SKIV2L2 Fusion Protein Ag3414 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |