missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 mouse anti-Human, Clone: 5B4G1, Proteintech
mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SLAM/CD150 |
---|---|
Clone | 5B4G1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836995
|
Proteintech
60043-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826995
|
Proteintech
60043-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD150, designated SLAM (Signaling Lymphocyte Activation Molecule) or CDw150, belongs to the immunoglobulin gene superfamily and is involved in T cell stimulation. SLAM is constitutively expressed on peripheral blood memory T cells, T cell clones, immature thymocytes, and a proportion of B cells, and is rapidly induced on naive T cells after activation. Activated B cells express the membrane bound form, the soluble form and the cytoplasmic isoforms of SLAM. It is suggested that signalling through homophilic SLAM-SLAM binding during B to B and B to T cell interactions enhances the expansion and differentiation of activated B cells. Reports suggest that the extracellular domain of CD150 is the receptor for the measles virus and acts as a co-activator on T cells and B cells.Specifications
SLAM/CD150 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD150, CDw150, IPO 3, SLAM, SLAMF1 | |
SLAMF1 | |
IgM | |
Caprylic Acid Precipitation | |
Antibody |
5B4G1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q13291 | |
6504 | |
SLAM/CD150 Fusion Protein Ag1386 | |
Primary | |
-20°C | |
SLAMF1 |