missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SLC22A23 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823415
|
Proteintech
24901-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813415
|
Proteintech
24901-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SLC22A23 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
A1A5C7, Q3UHH2, Q9QZG1 | |
63027, 64559, 73102 | |
SLC22A23 Fusion Protein Ag19036 | |
Primary | |
-20°C | |
SLC22A23 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BOCT2, C6orf85, SLC22A23 | |
SLC22A23 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |