missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18992-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the potassium-dependent sodium/calcium exchanger protein family. Alternative splicing results in multiple transcript variants.Specifications
SLC24A4 | |
Polyclonal | |
Unconjugated | |
SLC24A4 | |
NCKX4, SHEP6, SLC24A2, SLC24A4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123041, 238384 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CGQ8, Q8NFF2 | |
Slc24a4 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |