missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SLC24A4 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878804
|
Proteintech
18992-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868804
|
Proteintech
18992-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the potassium-dependent sodium/calcium exchanger protein family. Alternative splicing results in multiple transcript variants.Specifications
SLC24A4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8CGQ8, Q8NFF2 | |
123041, 238384 | |
Peptide | |
Primary | |
-20°C | |
SLC24A4 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NCKX4, SHEP6, SLC24A2, SLC24A4 | |
Slc24a4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |