missing translation for 'onlineSavingsMsg'
Learn More

SLC24A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27747-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825805

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The SLC24A5 gene is a member of the potassium-dependent sodium/calcium exchanger family and encodes an intracellular membrane protein with 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments. Sequence variation in this gene has been associated with differences in skin pigmentation.
Specifications

Specifications

SLC24A5
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q71RS6, Q8C261
JSX, NCKX5, SHEP4, SLC24A5
Rabbit
IgG
20 μL
-20°C
Primary
283652, 311387, 317750
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SLC24A5
Liquid
Slc24a5
SLC24A5 Fusion Protein Ag24254
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers