missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12253-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The oxoglutarate/malate carrier transports 2-oxoglutarate across the inner membranes of mitochondria in an electroneutral exchange for malate or other dicarboxylic acids (summary by Iacobazzi et al., 1992 ).Specifications
SLC25A11 | |
Polyclonal | |
Unconjugated | |
SLC25A11 | |
OGC, OGCP, SLC20A4, SLC25A11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8402 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q02978 | |
SLC25A11 | |
SLC25A11 Fusion Protein Ag2903 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |